Order Entry
Italy
ContactUsLinkComponent
6-(3,4-Dichlorophenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
6-(3,4-Dichlorophenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR32053-1G
Supplier:  Apollo Scientific
CAS Number:  
6-(3,4-Dichlorophenyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSOR32053-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.095 g/mol
Storage Temperature:Ambient
CAS Number:142499-56-7
Formula:C₁₂H₆Cl₂N₂O
MDL Number:MFCD00214977