Order Entry
Italy
ContactUsLinkComponent
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-1,4-dihydropyridin-4-one 95%
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-1,4-dihydropyridin-4-one 95%
Catalog # APOSOR32012-1G
Supplier:  Apollo Scientific
CAS Number:  
1-{4-[(2E)-3-(Dimethylamino)prop-2-enoyl]phenyl}-1,4-dihydropyridin-4-one 95%
Catalog # APOSOR32012-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.31 g/mol
Storage Temperature:Ambient
CAS Number:241488-20-0
Formula:C₁₆H₁₆N₂O₂
MDL Number:MFCD00139309