2-Chloro-7-methyl-7H-pyrrolo[2,3-d]pyrimidine
Catalog # APOSOR317013-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:167.596 g/mol
Storage Temperature:Refrigerator
CAS Number:1060816-67-2
Formula:C₇H₆ClN₃
MDL Number:MFCD13189420