
2,4-Dichloro-5-iodo-7H-pyrrolo[2,3-d]pyrimidine
Catalog # APOSOR317008-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:313.911 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1012785-51-1
Formula:C₆H₂Cl₂IN₃
MDL Number:MFCD13189364