Order Entry
Italy
ContactUsLinkComponent
2-Methoxy-6-methylpyrimidine-4-yl-amine 97%
2-Methoxy-6-methylpyrimidine-4-yl-amine 97%
Catalog # APOSOR315342-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-Methoxy-6-methylpyrimidine-4-yl-amine 97%
Catalog # APOSOR315342-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :139.155 g/mol
Storage Temperature:Ambient
CAS Number:51870-75-8
Formula:C₆H₉N₃O
MDL Number:MFCD00626432