


2-(4-Chlorophenyl)-2',4'-dichloroacetophenone
Catalog # APOSOR314002-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:299.58 g/mol
Melting Pt:65…66 °C
Storage Temperature:Ambient
UN:3077
ADR:9+EHS,III
:GHS05
:GHS07
:GHS09
CAS Number:94171-11-6
Formula:C₁₄H₉Cl₃O
MDL Number:MFCD18452152