Order Entry
Italy
ContactUsLinkComponent
Dodecanedioic acid monomethyl ester 97%
Irritant
Dodecanedioic acid monomethyl ester 97%
Catalog # APOSOR313077-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Dodecanedioic acid monomethyl ester 97%
Catalog # APOSOR313077-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :244.327 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:3903-40-0
Formula:C₁₃H₂₄O₄
MDL Number:MFCD00053323