Order Entry
Italy
ContactUsLinkComponent
5-Methyl-2-oxa-5,8-diazaspiro[3.5]nonane
Irritant
5-Methyl-2-oxa-5,8-diazaspiro[3.5]nonane
Catalog # APOSOR312278-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Methyl-2-oxa-5,8-diazaspiro[3.5]nonane
Catalog # APOSOR312278-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :142.199 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1369103-58-1
Formula:C₇H₁₄N₂O
MDL Number:MFCD22070771