Order Entry
Italy
ContactUsLinkComponent
Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate
Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate
Catalog # APOSOR311292-500MG
Supplier:  Apollo Scientific
CAS Number:  
Prop-2-en-1-yl 3,5-diamino-1H-pyrazole-4-carboxylate
Catalog # APOSOR311292-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :182.18 g/mol
Storage Temperature:Ambient
CAS Number:1613191-73-3
Formula:C₇H₁₀N₄O₂
MDL Number:MFCD27996731