Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-3-(4-chlorophenyl)-1,2,4-thiadiazole
5-Chloro-3-(4-chlorophenyl)-1,2,4-thiadiazole
5-Chloro-3-(4-chlorophenyl)-1,2,4-thiadiazole
APOSOR309416-1G
:
Apollo Scientific
:
5-Chloro-3-(4-chlorophenyl)-1,2,4-thiadiazole
APOSOR309416-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
231.102 g/mol
:
Ambient
:
887623-87-2
:
C₈H₄Cl₂N₂S
:
MFCD14707344