3,7-Dibromo-4-hydroxyquinoline
Catalog # APOSOR309323-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:302.95 g/mol
Storage Temperature:Ambient
CAS Number:1203579-53-6
Formula:C₉H₅Br₂NO
MDL Number:MFCD13192872