Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Aminomethyl)-1H-indazole
6-(Aminomethyl)-1H-indazole
6-(Aminomethyl)-1H-indazole
6-(Aminomethyl)-1H-indazole
APOSOR30834-250MG
:
Apollo Scientific
:
6-(Aminomethyl)-1H-indazole
APOSOR30834-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
147.177 g/mol
:
GHS07
:
Ambient
:
710943-26-3
:
C₈H₉N₃
:
MFCD09475774