
(3R)-3-(Methoxymethyl)morpholine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:218594-74-2
:
Formula:C₆H₁₃NO₂·ClH
MDL Number:MFCD09608106
:167.634 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together