
(2S)-2-(Aminomethyl)piperidine, N1-BOC protected
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:475105-35-2
:
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD06799358
:214.305 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together