6,8-Dimethyl-4-hydroxyquinoline
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:203626-58-8
- Formula:C₁₁H₁₁NO
- MDL Number:MFCD00272458
- Molecular Weight:173.211 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
- CAS Number:203626-58-8
- Formula:C₁₁H₁₁NO
- MDL Number:MFCD00272458
- Molecular Weight:173.211 g/mol
- Storage Temperature:Ambient



