Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N,N-Diethyl-1,4-phenylenediamine hydrochloride
N,N-Diethyl-1,4-phenylenediamine hydrochloride
N,N-Diethyl-1,4-phenylenediamine hydrochloride
APOSOR307789-5G
:
Apollo Scientific
:
N,N-Diethyl-1,4-phenylenediamine hydrochloride
APOSOR307789-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
200.708 g/mol
:
Ambient
:
2198-58-5
:
C₁₀H₁₆N₂·ClH
:
MFCD00035501