Order Entry
Italy
ContactUsLinkComponent
 
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-ethanol
Catalog # APOSOR307632-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(6,7-Diethoxy-1,2,3,4-tetrahydro-isoquinolin-1-yl)-ethanol
Catalog # APOSOR307632-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :265.348 g/mol
Storage Temperature:Ambient
CAS Number:955314-83-7
Formula:C₁₅H₂₃NO₃
MDL Number:MFCD07366621