6,7-Diethoxy-3,4-dihydro-1H-isoquinoline-1,2-dicarboxylic acid 2-tert-butyl ester
Catalog # APOSOR307617-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:365.421 g/mol
Storage Temperature:Ambient
CAS Number:1214645-14-3
Formula:C₁₉H₂₇NO₆
MDL Number:MFCD07366607