Order Entry
Italy
ContactUsLinkComponent
3-Iodo-4-methyl-1H-pyrazole
Irritant
3-Iodo-4-methyl-1H-pyrazole
Catalog # APOSOR307296-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Iodo-4-methyl-1H-pyrazole
Catalog # APOSOR307296-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :208 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:24086-18-8
Formula:C₄H₅IN₂
MDL Number:MFCD20230556