Order Entry
Italy
ContactUsLinkComponent
1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid
1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid
Catalog # APOSOR30727-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-(Tetrahydro-2H-pyran-2-yl)-1H-indazole-4-carboxylic acid
Catalog # APOSOR30727-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-(4-Carboxy-1H-indazol-1-yl)tetrahydro-2H-pyran, 4-Carboxy-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
 :
 :246.262 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1000576-23-7
Formula:C₁₃H₁₄N₂O₃
MDL Number:MFCD09870059