Order Entry
Italy
ContactUsLinkComponent
5-Chloro-1H-pyrazolo[4,3-d]pyrimidine
Irritant
5-Chloro-1H-pyrazolo[4,3-d]pyrimidine
Catalog # APOSOR305533-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Chloro-1H-pyrazolo[4,3-d]pyrimidine
Catalog # APOSOR305533-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :154.557 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:633328-98-0
Formula:C₅H₃ClN₄
MDL Number:MFCD18251003