
3-Chloro-1H-pyrazolo[3,4-c]pyridine
Catalog # APOSOR305367-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:153.569 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
CAS Number:76006-14-9
Formula:C₆H₄ClN₃
MDL Number:MFCD11846323