Order Entry
Italy
ContactUsLinkComponent
6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
Catalog # APOSOR305342-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine
Catalog # APOSOR305342-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :279.466 g/mol
Storage Temperature:Ambient
CAS Number:1260672-72-7
Formula:C₆H₃ClIN₃
MDL Number:MFCD18250995