Inserisci ordine
Italy
ContactUsLinkComponent
1-tert-Butyl 6-methyl 2,3-dihydro-1H-indole-1,6-dicarboxylate
1-tert-Butyl 6-methyl 2,3-dihydro-1H-indole-1,6-dicarboxylate
Catalog # APOSOR305249-250MG
Supplier:  Apollo Scientific
CAS Number:  
1-tert-Butyl 6-methyl 2,3-dihydro-1H-indole-1,6-dicarboxylate
Catalog # APOSOR305249-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Tipo di conf.:
    Bottle
  • Conf:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :277.316 g/mol
Storage Temperature:Ambiente
CAS Number:1220039-51-9
Formula:C₁₅H₁₉NO₄
MDL Number:MFCD09878912