Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-{2,7-Diazaspiro[3.5]nonan-2-yl}ethan-1-one
1-{2,7-Diazaspiro[3.5]nonan-2-yl}ethan-1-one
1-{2,7-Diazaspiro[3.5]nonan-2-yl}ethan-1-one
APOSOR304442-1G
:
Apollo Scientific
:
1-{2,7-Diazaspiro[3.5]nonan-2-yl}ethan-1-one
APOSOR304442-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
168.236 g/mol
:
Ambient
:
1474026-47-5
:
C₉H₁₆N₂O
:
MFCD28129054