Order Entry
Italy
ContactUsLinkComponent
1-{2,7-Diazaspiro[3.5]nonan-7-yl}ethan-1-one hydrochloride
1-{2,7-Diazaspiro[3.5]nonan-7-yl}ethan-1-one hydrochloride
Catalog # APOSOR304441-1G
Supplier:  Apollo Scientific
CAS Number:  
1-{2,7-Diazaspiro[3.5]nonan-7-yl}ethan-1-one hydrochloride
Catalog # APOSOR304441-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :204.697 g/mol
Storage Temperature:Ambient
CAS Number:2173992-33-9
Formula:C₉H₁₆N₂O·ClH
MDL Number:MFCD31617951