Order Entry
Italy
ContactUsLinkComponent
3-lambda-Thia-9-azaspiro[5.5]undecane-3,3-dione hydrochloride
3-lambda-Thia-9-azaspiro[5.5]undecane-3,3-dione hydrochloride
Catalog # APOSOR304434-1G
Supplier:  Apollo Scientific
CAS Number:  
3-lambda-Thia-9-azaspiro[5.5]undecane-3,3-dione hydrochloride
Catalog # APOSOR304434-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.763 g/mol
Storage Temperature:Ambient
CAS Number:1803582-31-1
Formula:C₉H₁₇NO₂S·ClH
MDL Number:MFCD28954306