4,7-Diazaspiro[2.5]octan-5-one
Catalog # APOSOR304394-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:126.156 g/mol
Storage Temperature:Ambient
CAS Number:1200114-14-2
Formula:C₆H₁₀N₂O
MDL Number:MFCD20526179