Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol 97%
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol 97%
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol 97%
APOSOR304004-1G
:
Apollo Scientific
:
(S)-(+)-2-Amino-1,1-diphenyl-1-propanol 97%
APOSOR304004-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
227.302 g/mol
:
100…102 °C
:
GHS07
:
Ambient
:
78603-91-5
:
C₁₅H₁₇NO
:
MFCD00145253