Order Entry
Italy
ContactUsLinkComponent
Ethyl 7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl 7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate
Catalog # APOSOR303911-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 7-amino-2-methylpyrazolo[1,5-a]pyrimidine-6-carboxylate
Catalog # APOSOR303911-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :220.228 g/mol
Melting Pt:192…194 °C
Storage Temperature:Ambient
CAS Number:2627-58-9
Formula:C₁₀H₁₂N₄O₂
MDL Number:MFCD03069208