
4-[2-(Morpholin-4-yl)ethyl]aniline
Catalog # APOSOR302403-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:206.284 g/mol
Melting Pt:92…94 °C
:GHS07
Storage Temperature:Ambient
CAS Number:262368-47-8
Formula:C₁₂H₁₈N₂O
MDL Number:MFCD04114508