4-(Azetidin-3-yl)morpholine hydrochloride
Catalog # APOSOR302316-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.66 g/mol
Storage Temperature:Ambient
CAS Number:223381-71-3
Formula:C₇H₁₄N₂O·ClH
MDL Number:MFCD11865371