Order Entry
Italy
ContactUsLinkComponent
But-3-en-1-amine, N-BOC protected
Irritant
Environment
But-3-en-1-amine, N-BOC protected
Catalog # APOSOR302285-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Environment
But-3-en-1-amine, N-BOC protected
Catalog # APOSOR302285-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :171.237 g/mol
Density:0.9293 g/cm³ (25 °C)
Flash Pt:101 °C
Storage Temperature:Refrigerator
UN:3082
ADR:9+EHS,III
 :GHS07
 :GHS09
CAS Number:156731-40-7
Formula:C₉H₁₇NO₂
MDL Number:MFCD11975862