Ethyl 2-hydroxyimidazo[1,2-a]pyridine-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27124-44-3
:
Formula:C₁₀H₁₀N₂O₃
MDL Number:MFCD26404011
:206.198 g/mol
Storage Temperature:Ambient
Frequently Bought Together