Order Entry
Italy
ContactUsLinkComponent
2,4-Dichloro-7,7-dimethyl-5,7-dihydrofuro[3,4-d]pyrimidine
2,4-Dichloro-7,7-dimethyl-5,7-dihydrofuro[3,4-d]pyrimidine
Catalog # APOSOR302091-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichloro-7,7-dimethyl-5,7-dihydrofuro[3,4-d]pyrimidine
Catalog # APOSOR302091-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.068 g/mol
Storage Temperature:Ambient
CAS Number:1260088-72-9
Formula:C₈H₈Cl₂N₂O
MDL Number:MFCD19688042