Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Methyl-1H-pyrazole-3-carboxamide
1-Methyl-1H-pyrazole-3-carboxamide 98%
1-Methyl-1H-pyrazole-3-carboxamide 98%
APOSOR301338-250MG
:
Apollo Scientific
:
1-Methyl-1H-pyrazole-3-carboxamide 98%
APOSOR301338-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
125.129 g/mol
:
Ambient
:
89179-62-4
:
C₅H₇N₃O
:
MFCD00464006