Order Entry
Italy
ContactUsLinkComponent
6-Bromo-1-methyl-1H-pyrrolo[3,2-b]pyridine 95+%
6-Bromo-1-methyl-1H-pyrrolo[3,2-b]pyridine 95+%
Catalog # APOSOR301261-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-Bromo-1-methyl-1H-pyrrolo[3,2-b]pyridine 95+%
Catalog # APOSOR301261-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :211.059 g/mol
Storage Temperature:Ambient
CAS Number:1086064-46-1
Formula:C₈H₇BrN₂
MDL Number:MFCD23703531