
2-Methyl-2-(methylamino)propan-1-ol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:27646-80-6
:
Formula:C₅H₁₃NO
MDL Number:MFCD12184529
:103.163 g/mol
Boiling Pt:162 °C
Density:0.878 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together