Order Entry
Italy
ContactUsLinkComponent
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[4,3-b]pyridine
Irritant
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[4,3-b]pyridine
Catalog # APOSOR300397-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazolo[4,3-b]pyridine
Catalog # APOSOR300397-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :282.136 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1416712-80-5
Formula:C₁₁H₁₂BrN₃O
MDL Number:MFCD22689847