Order Entry
Italy
ContactUsLinkComponent
6-Bromo-3-iodo-1H-pyrazolo[4,3-b]pyridine
Irritant
6-Bromo-3-iodo-1H-pyrazolo[4,3-b]pyridine
Catalog # APOSOR300391-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Bromo-3-iodo-1H-pyrazolo[4,3-b]pyridine
Catalog # APOSOR300391-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :323.917 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1305208-17-6
Formula:C₆H₃BrIN₃
MDL Number:MFCD20923312