
5-Bromo-2-cyclohexylisoindolin-1-one
Catalog # APOSOR300217-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:294.187 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:735351-81-2
Formula:C₁₄H₁₆BrNO
MDL Number:MFCD31561250