Order Entry
Italy
ContactUsLinkComponent
Ethyl 5-isopropyl-1-methyl-1H-pyrazole-3-carboxylate
Irritant
Ethyl 5-isopropyl-1-methyl-1H-pyrazole-3-carboxylate
Catalog # APOSOR300152-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 5-isopropyl-1-methyl-1H-pyrazole-3-carboxylate
Catalog # APOSOR300152-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :196.246 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1170483-34-7
Formula:C₁₀H₁₆N₂O₂
MDL Number:MFCD10699186