
3-Formyl-2,2,5,5-tetramethyl-1,3-thiazolidine-4-carboxylic acid
Catalog # APOSOR24610-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Melting Pt:179…180 °C
:217.285 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:55234-12-3
Formula:C₉H₁₅NO₃S
MDL Number:MFCD03939696