
Ethyl 1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylate
Catalog # APOSOR24264-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:219.24 g/mol
Melting Pt:87 °C
:GHS07
Storage Temperature:Ambient
CAS Number:175137-01-6
Formula:C₁₁H₁₃N₃O₂
MDL Number:MFCD00052539