
2-(Morpholin-4-yl)pyridine-3-sulphonyl chloride
Catalog # APOSOR23390-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:262.713 g/mol
Melting Pt:94…96 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
CAS Number:465514-17-4
Formula:C₉H₁₁ClN₂O₃S
MDL Number:MFCD03086194