Order Entry
Italy
ContactUsLinkComponent
4-Methyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid 97%
4-Methyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid 97%
Catalog # APOSOR23325-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Methyl-2-(pyrazin-2-yl)-1,3-thiazole-5-carboxylic acid 97%
Catalog # APOSOR23325-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :221.236 g/mol
Storage Temperature:Ambient
CAS Number:216959-92-1
Formula:C₉H₇N₃O₂S
MDL Number:MFCD00111662