Order Entry
Italy
ContactUsLinkComponent
1,9-Diazaspiro[5.5]undec-3-en-2-one hydrochloride 95+%
Irritant
1,9-Diazaspiro[5.5]undec-3-en-2-one hydrochloride 95+%
Catalog # APOSOR2325-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,9-Diazaspiro[5.5]undec-3-en-2-one hydrochloride 95+%
Catalog # APOSOR2325-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :202.681 g/mol
Melting Pt:>300 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1172814-19-5
Formula:C₉H₁₄N₂O·ClH
MDL Number:MFCD09998741