
5-(Pyridin-2-yl)thiophene-2-thiocarboxamide
Catalog # APOSOR22963-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:220.314 g/mol
Melting Pt:198…200 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:306934-91-8
Formula:C₁₀H₈N₂S₂
MDL Number:MFCD02677740