Order Entry
Italy
ContactUsLinkComponent
3-(4-Bromothien-2-yl)-1-(4-chlorophenyl)prop-2-en-1-one
Irritant
3-(4-Bromothien-2-yl)-1-(4-chlorophenyl)prop-2-en-1-one
Catalog # APOSOR21376-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(4-Bromothien-2-yl)-1-(4-chlorophenyl)prop-2-en-1-one
Catalog # APOSOR21376-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :327.624 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:219689-82-4
Formula:C₁₃H₈BrClOS
MDL Number:MFCD00117349