
Lithium 2-[(morpholin-4-yl)methyl]benzoate, Technical Grade
Catalog # APOSOR2079-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:227.185 g/mol
Melting Pt:>300 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:904696-60-2
Formula:C₁₂H₁₄NO₃·Li
MDL Number:MFCD08690277